accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNOSClON

CAS RN 515141-51-2

IUPAC Name: 6-(4-chlorophenyl)-3-{3-methoxy-4-[2-(pyrrolidin-1-yl)ethoxy]phenyl}-3H,4H-thieno[3,2-d]pyrimidin-4-one
Synonyms: 6-(4-chlorophenyl)-3-{3-methoxy-4-[2-(pyrrolidin-1-yl)ethoxy]phenyl}thieno[3,2-d]pyrimidin-4-one
Molecular Weight (g/mol): 482
Molecular Formula: C25H24ClN3O3S
InChi Key: MWULMTACIBZPGN-UHFFFAOYSA-N
SMILES: COC1=C(OCCN2CCCC2)C=CC(=C1)N1C=NC2=C(SC(=C2)C2=CC=C(Cl)C=C2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)