accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaNaOHNHNHOSOOOSOOOOSOOOSOOO(E)

CAS RN 51550-25-5

IUPAC Name: tetrasodium 4-hydroxy-5-{2-[(2E)-8-hydroxy-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}naphthalene-2,7-disulfonate
Synonyms: tetrasodium 4-hydroxy-5-{2-[(2E)-8-hydroxy-1-oxo-3,6-disulfonatonaphthalen-2-ylidene]hydrazin-1-yl}naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 738.5
Molecular Formula: C20H10N2Na4O15S4
InChi Key: CBWXTVSARCWSMJ-QNKYWQQSSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].OC1=CC(=CC2=CC(=CC(N\N=C3/C(=O)C4=C(C=C(C=C4O)S([O-])(=O)=O)C=C3S([O-])(=O)=O)=C12)S([O-])(=O)=O)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results