accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51939-71-0

IUPAC Name: 3,4-dihydro-2H-1-benzopyran-2-carboxylic acid
Synonyms: chroman-2-carboxylic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: SFLFCQJQOIZMHF-UHFFFAOYNA-N
SMILES: OC(=O)C1CCC2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results