Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromane-2-carboxylic acid, 97+%, Thermo Scientific™

Catalog No. RJC01246DE
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RJC01246DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RJC01246DE Supplier Thermo Scientific Chemicals Supplier No. RJC01246DE
Add to Cart

Chemical Identifiers

CAS 51939-71-0
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00044719
InChI Key SFLFCQJQOIZMHF-UHFFFAOYNA-N
Synonym chromane-2-carboxylic acid, chroman-2-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro, 2,3-dihydro-benzpyran-2-carboxylic acid, maybridge4_000177, 2-carboxychroman, chroman carboxylic acid, pubchem14568, 2-chroman-carboxylic acid
PubChem CID 2723665
IUPAC Name 3,4-dihydro-2H-chromene-2-carboxylic acid
SMILES OC(=O)C1CCC2=CC=CC=C2O1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 178.19
Percent Purity 97%
Chemical Name or Material Chromane-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.