accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHH2NNHOHSOHOO

CAS RN 52033-74-6

IUPAC Name: bis(phenylhydrazine); sulfuric acid
Synonyms: phenylhydrazinium sulphate (2:1)
Molecular Weight (g/mol): 314.36
Molecular Formula: C12H18N4O4S
InChi Key: GSKCKIQJTWVQLR-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NNC1=CC=CC=C1.NNC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results