accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH

CAS RN 52193-85-8

IUPAC Name: 1-(naphthalen-2-yl)ethan-1-ol
Synonyms: 1-(naphthalen-2-yl)ethanol
Molecular Weight (g/mol): 172.23
Molecular Formula: C12H12O
InChi Key: AXRKCRWZRKETCK-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results