accessibility menu, dialog, popup

UserName

Viewing profile as user:
HCHNONH2

CAS RN 5221-62-5

IUPAC Name: (prop-2-yn-1-yl)urea
Synonyms: prop-2-yn-1-ylurea
Molecular Weight (g/mol): 98.11
Molecular Formula: C4H6N2O
InChi Key: LJPYJRMMPVFEKR-UHFFFAOYSA-N
SMILES: NC(=O)NCC#C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)