Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propynylurea 98.0+%, TCI America™
SDP

Catalog No. P2171200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2171200MG 200 mg
P21711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2171200MG Supplier TCI America Supplier No. P2171200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5221-62-5
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.105
MDL Number MFCD00052817
InChI Key LJPYJRMMPVFEKR-UHFFFAOYSA-N
Synonym 1-prop-2-ynyl urea, 1-prop-2-yn-1-yl urea, n-prop-2-ynylurea, propargylurea, n-propargylurea, 2-propynylurea, prop-2-yn-1-yl urea, prop-2-yn-1-ylurea, prop-2-ynyl-urea, 1-prop-2-ynylurea
PubChem CID 2777182
IUPAC Name prop-2-ynylurea
SMILES C#CCNC(=O)N

Specifications

Melting Point 127°C
Quantity 200 mg
Formula Weight 98.11
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Propynylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.