Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Prop-2-ynylurea, Thermo Scientific™

Catalog No. CD11100DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CD11100DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CD11100DA Supplier Thermo Scientific Chemicals Supplier No. CD11100DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5221-62-5
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.105
MDL Number MFCD00052817
InChI Key LJPYJRMMPVFEKR-UHFFFAOYSA-N
Synonym 1-prop-2-ynyl urea, 1-prop-2-yn-1-yl urea, n-prop-2-ynylurea, propargylurea, n-propargylurea, 2-propynylurea, prop-2-yn-1-yl urea, prop-2-yn-1-ylurea, prop-2-ynyl-urea, 1-prop-2-ynylurea
PubChem CID 2777182
IUPAC Name prop-2-ynylurea
SMILES C#CCNC(=O)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula H2NC(O)NHCH2Cidentical toCH
Quantity 1 g
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material N-Prop-2-ynylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.