accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 525-05-3

IUPAC Name: disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate
Synonyms: nitroso-R salt
Molecular Weight (g/mol): 377.25
Molecular Formula: C10H5NNa2O8S2
InChi Key: DMKMTGULLYISBH-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OC1=C(N=O)C2=CC=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (4)
  • (4)
  • (4)
Physical Form 
  • (7)
Percent Purity 
  • (3)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (8)
  • (4)
  • (4)
  • (4)
  • (7)
  • (3)
  • (1)
  • (2)
  • (3)