Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt, 95%, Thermo Scientific™

Catalog No. AC416191000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC416191000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC416191000 Supplier Thermo Scientific Chemicals Supplier No. 416191000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 525-05-3
Molecular Formula C10H5NNa2O8S2
Molecular Weight (g/mol) 377.25
MDL Number MFCD00149246
InChI Key DMKMTGULLYISBH-UHFFFAOYSA-L
Synonym nitroso-r salt, nitroso r, sodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate, disodium 1-nitroso-2-naphthol-3,6-disulfonate, nitroso r salt, 1-nitroso-3,6-disulfonate, 2,7-naphthalenedisulfonic acid, 3-hydroxy-4-nitroso-, disodium salt, 3-hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt, 1-nitroso-2-naphthol-3,6-disulfonic acid disodium salt, disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulphonate
PubChem CID 10672
IUPAC Name disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].OC1=C(N=O)C2=CC=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown-Yellow to Orange-Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Quantity 100 g
Beilstein 11, II, 190
Merck Index 15, 6731
Formula Weight 377.25
Physical Form Crystalline Powder
Chemical Name or Material 1-Nitroso-2-naphthol-3, 6-disulfonic acid disodium salt, pract., 95%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.