Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-Nitroso-2,7- Naphthalenedisulfonic Acid Disodium Salt, Solstice

Catalog No. 60047460 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-460 25 g Glass bottle
60-047-459 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-460 Supplier Solstice Supplier No. 7395025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for spectrophotometric det. of Co, Fe, K, Ag, Ba, Ca, Co, Fe, Ni, Pb

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 525-05-3
Molecular Formula C10H5NNa2O8S2
Molecular Weight (g/mol) 377.25
MDL Number MFCD00149246
InChI Key DMKMTGULLYISBH-UHFFFAOYSA-L
Synonym nitroso-r salt, nitroso r, sodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate, disodium 1-nitroso-2-naphthol-3,6-disulfonate, nitroso r salt, 1-nitroso-3,6-disulfonate, 2,7-naphthalenedisulfonic acid, 3-hydroxy-4-nitroso-, disodium salt, 3-hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt, 1-nitroso-2-naphthol-3,6-disulfonic acid disodium salt, disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulphonate
PubChem CID 10672
IUPAC Name disodium;3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].OC1=C(N=O)C2=CC=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Melting Point 300°C
Color Yellow
Packaging Glass bottle
Quantity 25 g
UN Number NONH for all modes of transport
Beilstein 3643930
Formula Weight 377.26g/mol
Chemical Name or Material 3-Hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt
Hazard Statement P280
RTECSNumber 208-369-1
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.