accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOOOOO

CAS RN 52522-49-3

IUPAC Name: 3,4-bis(benzyloxy)-5-[(benzyloxy)methyl]oxolan-2-yl 4-nitrobenzoate
Synonyms: 3,4-bis(benzyloxy)-5-[(benzyloxy)methyl]oxolan-2-yl 4-nitrobenzoate
Molecular Weight (g/mol): 569.61
Molecular Formula: C33H31NO8
InChi Key: VBDBDZHLJKDSSB-UHFFFAOYNA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C(=O)OC1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results