accessibility menu, dialog, popup

UserName

Viewing profile as user:
RhHIIHCOHCCCCC

CAS RN 5256-79-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 447.81
Molecular Formula: C6H3I2ORh
InChi Key: —
SMILES: [Rh].I.I.[C-]#[O+].[c-]1[cH-][cH-][cH-][cH-]1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)