accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH(S)HNOHO

CAS RN 526-31-8

IUPAC Name: (2S)-3-(1H-indol-3-yl)-2-(methylamino)propanoic acid
Synonyms: N-methyl-L-tryptophan N-metil-L-triptofano
Molecular Weight (g/mol): 218.26
Molecular Formula: C12H14N2O2
InChi Key: CZCIKBSVHDNIDH-NSHDSACASA-N
SMILES: CN[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)