CAS RN 527-04-8
IUPAC Name:
2,3,4,5-tetrahydroxyhexanedioic acid diamine
Synonyms:
D-saccharate diamine
Molecular Weight (g/mol):
244.2
Molecular Formula:
C6H16N2O8
InChi Key:
LZWIOJIGSVGZID-UHFFFAOYNA-N
SMILES:
N.N.OC(C(O)C(O)C(O)=O)C(O)C(O)=O
1
–
1
of
1
results
1
MP Biomedicals, Inc Ammonium Mucate, MP Biomedicals
CAS: 527-04-8 Molecular Formula: C6H18N2O8+2 Molecular Weight (g/mol): 246.216 InChI Key: LZWIOJIGSVGZID-SXKXKJGMSA-P Synonym: galactaric acid diammonium salt PubChem CID: 101318330 IUPAC Name: diazanium;(2S,3R,4S,5R)-2,3,4,5-tetrahydroxyhexanedioic acid SMILES: C(C(C(C(=O)O)O)O)(C(C(=O)O)O)O.[NH4+].[NH4+]