accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 527-53-7

IUPAC Name: 1,2,3,5-tetramethylbenzene
Synonyms: 1,2,3,5-tetramethylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: BFIMMTCNYPIMRN-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)C(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)