Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,5-Tetramethylbenzene, Mixture with Durene 80%, Thermo Scientific™

Catalog No. AC167195000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC167195000 500 g
AC167191000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167195000 Supplier Thermo Scientific Chemicals Supplier No. 167195000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-53-7
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00008524
InChI Key BFIMMTCNYPIMRN-UHFFFAOYSA-N
Synonym isodurene, benzene, 1,2,3,5-tetramethyl, unii-0jbi5y5a5z, 1,3,4,5-tetramethylbenzene, 0jbi5y5a5z, ccris 8661, acmc-1avx4, dsstox_cid_6119, 1,3,5-tetramethylbenzene
PubChem CID 10695
IUPAC Name 1,2,3,5-tetramethylbenzene
SMILES CC1=CC(C)=C(C)C(C)=C1

Specifications

CAS Min % 75.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.89
Infrared Spectrum Authentic
Refractive Index 1.5110 to 1.5130 (20°C, 589nm)
Linear Formula C6H2(CH3)4
Quantity 500 g
Beilstein 05, 430
Merck Index 15, 5211
Formula Weight 134.22
Percent Purity ≥75%
Physical Form Liquid
Chemical Name or Material 1,2,3,5-Tetramethylbenzene, mixture with durene, 80%
Show More Show Less
EINECSNumber 208-417-1
RTECSNumber DC0475000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.