Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,5-Tetramethylbenzene 70.0+%, TCI America™
SDP

Catalog No. T05655ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T05655ML 5 mL
T056525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T05655ML Supplier TCI America Supplier No. T05655ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 527-53-7
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00008524
InChI Key BFIMMTCNYPIMRN-UHFFFAOYSA-N
Synonym isodurene, benzene, 1,2,3,5-tetramethyl, unii-0jbi5y5a5z, 1,3,4,5-tetramethylbenzene, 0jbi5y5a5z, ccris 8661, acmc-1avx4, dsstox_cid_6119, 1,3,5-tetramethylbenzene
PubChem CID 10695
IUPAC Name 1,2,3,5-tetramethylbenzene
SMILES CC1=CC(C)=C(C)C(C)=C1

Specifications

Color Yellow
Boiling Point 198°C
Quantity 5 mL
Formula Weight 134.22
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3,5-Tetramethylbenzene
EINECSNumber (3)-0007
RTECSNumber DC0475000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.