accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3NN

CAS RN 5270-94-0

IUPAC Name: 4,6-dimethoxypyrimidine
Synonyms: 4,6-dimethoxypyrimidine
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: FPSPPRZKBUVEJQ-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=NC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results