Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dimethoxypyrimidine 98.0+%, TCI America™
SDP

Catalog No. D29595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29595G Supplier TCI America Supplier No. D29595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5270-94-0
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00955844
InChI Key FPSPPRZKBUVEJQ-UHFFFAOYSA-N
PubChem CID 259821
IUPAC Name 4,6-dimethoxypyrimidine
SMILES COC1=CC(=NC=N1)OC

Specifications

Melting Point 33°C
Color White
Boiling Point 186°C
Quantity 5 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4,6-Dimethoxypyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.