accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOONNSOOO

CAS RN 52746-49-3

IUPAC Name: disodium 4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzene-1-sulfonate
Synonyms: disodium 4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate
Molecular Weight (g/mol): 536.48
Molecular Formula: C24H14N2Na2O6S2
InChi Key: PCNDSIWXTYFWIA-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (7)
Quantity 
  • (3)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (5)
  • (1)
  • (2)
  • (1)
  • (7)
  • (3)
  • (1)
  • (4)
  • (3)