Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bathophenanthrolinedisulfonic Acid Disodium Salt Hydrate, Sigma-Aldrich

Catalog No. 01259011 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-011 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-011 Supplier MilliporeSigma Supelco Supplier No. B13755G
Add to Cart
Add to Cart

Bathophenanthrolinedisulfonic acid is a metal chelator. It permits iron determination to be made in aqueous medium without need for solvent extraction. It forms a complex with Fe2+ and makes it unavailable to Fe2+transporter, thereby inhibiting the transport of iron into chloroplasts. Bathophenanthrolinedisulfonic acid is also used in the assay for determining intracellular glutathione.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52746-49-3
Molecular Weight (g/mol) 536.49 g/mol

Specifications

Quantity 5 g
Percent Purity ≥95%
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.