accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2N(S)ClClHOO

CAS RN 52794-99-7

IUPAC Name: (2S)-2-amino-3-(3,4-dichlorophenyl)propanoic acid
Synonyms: (2S)-2-amino-3-(3,4-dichlorophenyl)propanoic acid
Molecular Weight (g/mol): 234.08
Molecular Formula: C9H9Cl2NO2
InChi Key: NRCSJHVDTAAISV-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=C(Cl)C(Cl)=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results