accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOSClNOO

CAS RN 528-76-7

IUPAC Name: [(2,4-dinitrophenyl)sulfanyl]chlorane
Synonyms: [(2,4-dinitrophenyl)sulfanyl]chlorane
Molecular Weight (g/mol): 234.61
Molecular Formula: C6H3ClN2O4S
InChi Key: GPXDNWQSQHFKRB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(SCl)C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results