CAS RN 529-38-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Cocaethylene Solution, 1.0 mg/mL in acetonitrile, Sold by MilliporeSigma™ Supelco™
A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for GC/MS or LC/MS methods in clinical toxicology, forensic analysis, or urine drug testing.