accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3OOCH3OOCH3OOOOCH3OOCH3OOCH3OOCH3OH3COOOOCH3OOCH3OOCH3OOOOCH3OOCH3OOCH3OOCH3OHOOOHOOHOHOHOOHHOHOHOOOHOOHOHOHOOHHOHOHOOOOOOOOHOHOOOOHOOOHOOOHOOOHOOOOOHOOOHOOOOHOHOOOOHOOOHOOOOHOOHOHOO

CAS RN 52907-01-4

IUPAC Name: bis(2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol); 4-({[4,5,6-tris(2,4-dicarboxybenzoyloxy)-3-{[3,4,5-tris(2,4-dicarboxybenzoyloxy)-6-[(2,4-dicarboxybenzoyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methoxy}carbonyl)benzene-1,3-dicarboxylic acid; bis([4,5,6-tris(acetyloxy)-3-{[3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methyl acetate)
Synonyms: 4-({[4,5,6-tris(2,4-dicarboxybenzoyloxy)-3-{[3,4,5-tris(2,4-dicarboxybenzoyloxy)-6-[(2,4-dicarboxybenzoyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methoxy}carbonyl)benzene-1,3-dicarboxylic acid; bis([4,5,6-tris(acetyloxy)-3-{[3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methyl acetate); bis(starch)
Molecular Weight (g/mol): 3921.09
Molecular Formula: C164H174O111
InChi Key: WZNRVWBKYDHTKI-UHFFFAOYNA-N
SMILES: OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O.OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O.CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O.CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O.OC(=O)C1=CC=C(C(=O)OCC2OC(OC(=O)C3=CC=C(C=C3C(O)=O)C(O)=O)C(OC(=O)C3=CC=C(C=C3C(O)=O)C(O)=O)C(OC(=O)C3=CC=C(C=C3C(O)=O)C(O)=O)C2OC2OC(COC(=O)C3=CC=C(C=C3C(O)=O)C(O)=O)C(OC(=O)C3=CC=C(C=C3C(O)=O)C(O)=O)C(OC(=O)C3=CC=C(C=C3C(O)=O)C(O)=O)C2OC(=O)C2=CC=C(C=C2C(O)=O)C(O)=O)C(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results