accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHO

CAS RN 52938-97-3

IUPAC Name: 5-phenylfuran-2-carboxylic acid
Synonyms: 5-phenylfuran-2-carboxylic acid
Molecular Weight (g/mol): 188.18
Molecular Formula: C11H8O3
InChi Key: GUOMINFEASCICM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)