accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5298-72-6

IUPAC Name: 1-(1-methylpiperidin-2-yl)methanamine
Synonyms: 1-(1-methylpiperidin-2-yl)methanamine
Molecular Weight (g/mol): 128.22
Molecular Formula: C7H16N2
InChi Key: PPUMJZMVFCLQBI-UHFFFAOYNA-N
SMILES: CN1CCCCC1CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results