CAS RN 5299-57-0
IUPAC Name:
ethenyl undec-10-enoate
Synonyms:
ethenyl undec-10-enoate
Molecular Weight (g/mol):
210.32
Molecular Formula:
C13H22O2
InChi Key:
HUGGPHJJSYXCDJ-UHFFFAOYSA-N
SMILES:
C=CCCCCCCCCC(=O)OC=C
1
–
1
of
1
results
1
Vinyl 10-Undecenoate (stabilized with MEHQ) 92.0+%, TCI America™
CAS: 5299-57-0 Molecular Formula: C13H22O2 Molecular Weight (g/mol): 210.32 MDL Number: MFCD00142351 InChI Key: HUGGPHJJSYXCDJ-UHFFFAOYSA-N Synonym: 10-Undecenoic Acid Vinyl Ester PubChem CID: 544393 IUPAC Name: ethenyl undec-10-enoate SMILES: C=CCCCCCCCCC(=O)OC=C