accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)OOOH3CCH3NOH

CAS RN 53034-38-1

IUPAC Name: 15-[(2R)-2-ethyl-3-hydroxy-4,4-dimethyl-1,3-oxazolidin-2-yl]pentadecanoate
Synonyms: 15-[(2R)-2-ethyl-3-hydroxy-4,4-dimethyl-1,3-oxazolidin-2-yl]pentadecanoate
Molecular Weight (g/mol): 384.58
Molecular Formula: C22H42NO4
InChi Key: BPDFMDXURXAITH-JOCHJYFZSA-M
SMILES: CC[C@]1(CCCCCCCCCCCCCCC([O-])=O)OCC(C)(C)N1O
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)