Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

16-DOXYL-stearic Acid Free Radical 97.0+%, TCI America™
SDP

Catalog No. D239925MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D239925MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D239925MG Supplier TCI America Supplier No. D239925MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53034-38-1
Molecular Formula C22H42NO4
Molecular Weight (g/mol) 384.58
MDL Number MFCD00010066
InChI Key BPDFMDXURXAITH-JOCHJYFZSA-M
Synonym 16-doxyl-stearic acid, 16-nitroxystearate, 16-doxylstearic acid, 16-doxyl-stearic acid, free radical, 16-doxylstearate, 16-doxyl-ste, 16-doxystearic acid, fasl 1,14, 16doxyl, 15-2-ethyl-3
PubChem CID 2724321
IUPAC Name 15-[(2R)-2-ethyl-3-hydroxy-4,4-dimethyl-1,3-oxazolidin-2-yl]pentadecanoate
SMILES CC[C@]1(CCCCCCCCCCCCCCC([O-])=O)OCC(C)(C)N1O

Specifications

Melting Point 58°C
Color Yellow
Quantity 25 mg
Formula Weight 384.58
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 16-DOXYL-stearic Acid Free Radical
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.