Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

16-DOXYL-stearic Acid, Free Radical, Thermo Scientific™

Catalog No. AC225120250
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC225120250 25 mg
AC225120100 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC225120250 Supplier Thermo Scientific Chemicals Supplier No. 225120250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53034-38-1
Molecular Formula C22H42NO4
Molecular Weight (g/mol) 384.58
MDL Number MFCD00010066
InChI Key BPDFMDXURXAITH-JOCHJYFZSA-M
Synonym 16-doxyl-stearic acid, 16-nitroxystearate, 16-doxylstearic acid, 16-doxyl-stearic acid, free radical, 16-doxylstearate, 16-doxyl-ste, 16-doxystearic acid, fasl 1,14, 16doxyl, 15-2-ethyl-3
PubChem CID 2724321
IUPAC Name 15-[(2R)-2-ethyl-3-hydroxy-4,4-dimethyl-1,3-oxazolidin-2-yl]pentadecanoate
SMILES CC[C@]1(CCCCCCCCCCCCCCC([O-])=O)OCC(C)(C)N1O

Specifications

Quantity 25 mg
Formula Weight 384.57
Chemical Name or Material 16-DOXYL-stearic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.