accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClCl

CAS RN 5312-73-2

IUPAC Name: 1-(dichloromethoxy)butane
Synonyms: 1-(dichloromethoxy)butane
Molecular Weight (g/mol): 157.03
Molecular Formula: C5H10Cl2O
InChi Key: GVVHBNABFJZWIC-UHFFFAOYSA-N
SMILES: CCCCOC(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results