Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Dichloromethyl Ether 98.0+%, TCI America™
SDP

Catalog No. D227810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D227810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D227810G Supplier TCI America Supplier No. D227810G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5312-73-2
Molecular Formula C5H10Cl2O
Molecular Weight (g/mol) 157.03
MDL Number MFCD00191420
InChI Key GVVHBNABFJZWIC-UHFFFAOYSA-N
Synonym Dichloromethyl Butyl Ether
PubChem CID 4278354
IUPAC Name 1-(dichloromethoxy)butane
SMILES CCCCOC(Cl)Cl

Specifications

Color Colorless
Boiling Point 50°C
Quantity 10 g
UN Number 1993
Formula Weight 157.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Dichloromethyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.