accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5314-37-4

IUPAC Name: sodium [1,1'-biphenyl]-4,4'-disulfonic acid
Synonyms: sodium [1,1'-biphenyl]-4,4'-disulfonic acid
Molecular Weight (g/mol): 337.32
Molecular Formula: C12H10NaO6S2
InChi Key: HGAPLVLNYHEDFN-UHFFFAOYSA-N
SMILES: [Na+].OS(=O)(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results