accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOOCH3CH3

CAS RN 53148-32-6

IUPAC Name: (4-{[(2-ethylhexanoyl)oxy]methyl}cyclohexyl)methyl 2-ethylhexanoate
Synonyms: (4-{[(2-ethylhexanoyl)oxy]methyl}cyclohexyl)methyl 2-ethylhexanoate
Molecular Weight (g/mol): 396.61
Molecular Formula: C24H44O4
InChi Key: WZCWEBVKSPNNNN-UHFFFAOYNA-N
SMILES: CCCCC(CC)C(=O)OCC1CCC(COC(=O)C(CC)CCCC)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results