accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5318-27-4

NH2HN

CAS RN 5318-27-4

IUPAC Name: 1H-indol-6-amine
Synonyms: 1H-indol-6-amine
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: MIMYTSWNVBMNRH-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=CNC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)