CAS RN 532-27-4
CAS RN 532-27-4
IUPAC Name:
2-chloro-1-phenylethan-1-one
Synonyms:
phenacyl chloride
Molecular Weight (g/mol):
154.59
Molecular Formula:
C8H7ClO
InChi Key:
IMACFCSSMIZSPP-UHFFFAOYSA-N
SMILES:
ClCC(=O)C1=CC=CC=C1
1
–
2
of
2
results
1
2-Chloroacetophenone, 98%, Thermo Scientific Chemicals
CAS: 532-27-4 Molecular Formula: C8H7ClO Molecular Weight (g/mol): 154.59 MDL Number: MFCD00000933 InChI Key: IMACFCSSMIZSPP-UHFFFAOYSA-N Synonym: 2-chloroacetophenone,phenacyl chloride,omega-chloroacetophenone,chloroacetophenone,tear gas,mace lacrimator,alpha-chloroacetophenone,ethanone, 2-chloro-1-phenyl,mace,1-chloroacetophenone PubChem CID: 10757 IUPAC Name: 2-chloro-1-phenylethan-1-one SMILES: ClCC(=O)C1=CC=CC=C1
2
2-Chloroacetophenone, SPEX CertiPrep™
CAS: 532-27-4 Molecular Formula: C8H7ClO Molecular Weight (g/mol): 154.59 MDL Number: MFCD00000933 InChI Key: IMACFCSSMIZSPP-UHFFFAOYSA-N PubChem CID: 10757 IUPAC Name: 2-chloro-1-phenylethan-1-one SMILES: ClCC(=O)C1=CC=CC=C1