accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CHOOO(Z)

CAS RN 5323-95-5

IUPAC Name: sodium (9Z)-12-hydroxyoctadec-9-enoate
Synonyms: sodium ricinoleate
Molecular Weight (g/mol): 320.45
Molecular Formula: C18H33NaO3
InChi Key: IJRHDFLHUATAOS-MWMYENNMNA-M
SMILES: [Na+].CCCCCCC(O)C\C=C/CCCCCCCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results