accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOHSOOSOHOO

CAS RN 5325-43-9

IUPAC Name: ethane-1,2-disulfonic acid sodium
Synonyms: ethanedisulfonic acid sodium
Molecular Weight (g/mol): 213.17
Molecular Formula: C2H6NaO6S2
InChi Key: XTZUFSDRWFDYGH-UHFFFAOYSA-N
SMILES: [Na].OS(=O)(=O)CCS(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results