Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 1,2-Ethanedisulfonate 98.0+%, TCI America™
SDP

Catalog No. E00315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E00315G 5 g
E003125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E00315G Supplier TCI America Supplier No. E00315G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5325-43-9
Molecular Formula C2H4Na2O6S2
Molecular Weight (g/mol) 234.148
MDL Number MFCD00024998
InChI Key BJTHMUJCKBTCFR-UHFFFAOYSA-L
Synonym sodium ethane-1,2-disulfonate, 1,2-ethanedisulfonic acid disodium salt, disodium 1,2-ethanedisulfonate, 1,2-ethanedisulfonic acid, disodium salt, unii-2yzu14uifp, 2yzu14uifp, disodium ethanedisulfonic acid, disodium ethane-1,2-disulfonate, 1,2-ethanedisulfonic acid, sodium salt 1:2, sodium 1,2-ethanedisulfonate
PubChem CID 79220
IUPAC Name disodium;ethane-1,2-disulfonate
SMILES C(CS(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 234.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 1,2-Ethanedisulfonate
EINECSNumber (2)-1632
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.