accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBrCH3

CAS RN 533-68-6

IUPAC Name: ethyl 2-bromobutanoate
Synonyms: ethyl 2-bromobutanoate
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H11BrO2
InChi Key: XIMFCGSNSKXPBO-UHFFFAOYNA-N
SMILES: CCOC(=O)C(Br)CC
Boiling Point 
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)