Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Bromobutyrate 98.0+%, TCI America™
SDP

Catalog No. B0800500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0800500G 500 g
B080025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0800500G Supplier TCI America Supplier No. B0800500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 533-68-6
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000155
InChI Key XIMFCGSNSKXPBO-UHFFFAOYNA-N
Synonym ethyl 2-bromobutyrate, dl-ethyl 2-bromobutyrate, 2-bromobutanoic acid ethyl ester, butanoic acid, 2-bromo-, ethyl ester, 2-bromobutyric acid ethyl ester, ethyl .alpha.-bromobutyrate, 2-bromo-butyric acid ethyl ester, butyric acid, 2-bromo-, ethyl ester, 2-bromo-butanoicaciethylester, ethyl alpha-bromobutyrate
PubChem CID 79039
IUPAC Name ethyl 2-bromobutanoate
SMILES CCOC(=O)C(Br)CC

Specifications

Color Yellow
Boiling Point 178°C
Quantity 500 g
Formula Weight 195.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Bromobutyrate
EINECSNumber (2)-1179
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.