CAS RN 5332-73-0
IUPAC Name:
3-methoxypropan-1-amine
Synonyms:
3-methoxypropylamine
Molecular Weight (g/mol):
89.14
Molecular Formula:
C4H11NO
InChi Key:
FAXDZWQIWUSWJH-UHFFFAOYSA-N
SMILES:
COCCCN
1
–
6
of
6
results
1
Promotions Available
3-Methoxypropylamine, 99+%, Thermo Scientific Chemicals
3-Methoxypropylamine, 99+%, Thermo Scientific Chemicals
2
Promotions Available
3-Methoxypropylamine, 99%
3-Methoxypropylamine, 99%
3
Promotions Available
Thermo Scientific™ 3-Methoxypropylamine, 99%