accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NH3CCH3NCH3H3C

CAS RN 53369-79-2

IUPAC Name: [3-(dimethylamino)-2,2-dimethylpropyl]dimethylamine
Synonyms: [3-(dimethylamino)-2,2-dimethylpropyl]dimethylamine
Molecular Weight (g/mol): 158.29
Molecular Formula: C9H22N2
InChi Key: MOBAUGYLXJCSAW-UHFFFAOYSA-N
SMILES: CN(C)CC(C)(C)CN(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results