CAS RN 53398-86-0
IUPAC Name:
(2E)-hex-2-en-1-yl hexanoate
Synonyms:
trans-2-hexenyl hexanoate
Molecular Weight (g/mol):
198.31
Molecular Formula:
C12H22O2
InChi Key:
UQPLEMTXCSYMEK-VQHVLOKHSA-N
SMILES:
CCCCCC(=O)OC\C=C\CCC
1
–
1
of
1
results
1
trans-2-Hexenyl Hexanoate 97.0+%, TCI America™
CAS: 53398-86-0 Molecular Formula: C12H22O2 Molecular Weight (g/mol): 198.31 MDL Number: MFCD00036551 InChI Key: UQPLEMTXCSYMEK-VQHVLOKHSA-N Synonym: Hexanoic Acid trans-2-Hexenyl Ester PubChem CID: 5352973 IUPAC Name: (2E)-hex-2-en-1-yl hexanoate SMILES: CCCCCC(=O)OC\C=C\CCC