accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53491-30-8

H3COCl

CAS RN 53491-30-8

IUPAC Name: (2-chloropropoxy)benzene
Synonyms: (2-chloropropoxy)benzene
Molecular Weight (g/mol): 170.64
Molecular Formula: C9H11ClO
InChi Key: LTRAIWUUCBYXHZ-UHFFFAOYNA-N
SMILES: CC(Cl)COC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results