accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HNONH2

CAS RN 5351-17-7

IUPAC Name: 4-aminobenzohydrazide
Synonyms: 4-aminobenzoylhydrazine
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: WPBZMCGPFHZRHJ-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)
Form 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)