Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobenzhydrazide, 98%, Thermo Scientific™

Catalog No. AAA1096322
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1096322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1096322 Supplier Thermo Scientific Chemicals Supplier No. A1096322

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Aminobenzhydrazide has an important role in the peroxidation activity of MPO. It is also used in enzyme preparations and in cell cultures.

Solubility
Soluble in DMSO (10 mg/ml), dilute acids, and water (partly miscible).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5351-17-7
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.169
MDL Number MFCD00007606
InChI Key WPBZMCGPFHZRHJ-UHFFFAOYSA-N
Synonym 4-aminobenzhydrazide, amben hydrazide, p-aminobenzhydrazide, 4-aminobenzoic acid hydrazide, aminostimil, 4-aminobenzoylhydrazine, p-aminobenzoic acid hydrazide, p-aminobenzoyl hydrazide, benzoic acid, 4-amino-, hydrazide, p-aminobenzoic hydrazide
PubChem CID 21450
IUPAC Name 4-aminobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)N

Specifications

Melting Point 225°C to 230°C
Quantity 100 g
Beilstein 639053
Solubility Information Soluble in DMSO (10mg/ml),dilute acids,and water (partly miscible).
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material 4-Aminobenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-324-4
RTECSNumber DG2580000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.